Merck
  • Levels, sources and health risks of carbonyls and BTEX in the ambient air of Beijing, China.
Levels, sources and health risks of carbonyls and BTEX in the ambient air of Beijing, China.
Journal of environmental sciences (China) (2012-07-13)
Yujie Zhang, Yujing Mu, Junfeng Liu, Abdelwahid Mellouki
ABSTRACT
The atmospheric concentrations of carbonyls and BTEX (benzene, toluene, ethylbenzene, m,p-xylene and o-xylene) were measured simultaneously at a same sampling site in Beijing from September 2008 to August 2010. The average concentrations of the total measured carbonyls during autumn, winter, spring, and summer were 37.7, 31.3, 39.7, 50.5 microg/m3, respectively, and maximal values for their diurnal variations usually happened at noontime. In contrast to carbonyls, the average concentrations of the total measured BTEX during the four seasons were 27.2, 31.9, 23.2, 19.1 microg/m3, respectively, and minimal values for their diurnal variations always occurred in the early afternoon. The average concentration for carbonyls increased about 24% from September 2008-August 2009 to September 2009-August 2010, for BTEX, increased about 15%. Integrated life time cancer risks for three carcinogens (benzene, formaldehyde and acetaldehyde) in Beijing exceeded the value of 1E-06, and the hazard quotient (HQ) of non-cancer risk of exposure to formaldehyde exceeded unity.
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 1% in chloroform-d (99.8 atom % D)
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 10% in chloroform-d (99.8 atom % D), NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 5% in chloroform-d (99.8 atom % D), TMS 1 %, NMR tube size 5 mm × 7 in.
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.1 %
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 10% in chloroform-d (99.8 atom % D)
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d ("100%", 99.96 atom % D)
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 8 mm × 8 in.
Supelco
m-Xylene, analytical standard
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 5% in chloroform-d (99.8 atom % D), TMS 2 %, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in. , ultra-thin wall
Supelco
m-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), chromium(III) acetylacetonate 0.1 mg/mL, TMS 0.01 %, NMR tube size 5 mm × 8 in.
Supelco
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in.
Supelco
Ethylbenzene solution, suitable for NMR (reference standard), 10% in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in. , ultra-thin wall
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
p-Xylene, ReagentPlus®, 99%
Sigma-Aldrich
Xylenes, ACS reagent, ≥98.5% xylenes + ethylbenzene basis
Sigma-Aldrich
Ethylbenzene, ReagentPlus®, 99%
Sigma-Aldrich
Xylenes, histological grade
Sigma-Aldrich
p-Xylene, suitable for HPLC, ≥99%
Sigma-Aldrich
p-Xylene, anhydrous, ≥99%
Supelco
Ethylbenzene, analytical standard
Sigma-Aldrich
Xylenes, reagent grade
Sigma-Aldrich
Ethylbenzene, anhydrous, 99.8%
Supelco
p-Xylene, analytical standard
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
p-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material