Merck
[Chemical constituents from stems of Dysoxylum laxiracemosum].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2012-07-19)
Ting Tang, Laifu Zuo, Zhi Na, Youkai Xu
ABSTRACT
Twelve compounds were separated from stems of Dysoxylum laxiracemosum and their structures were identified by spectrum analysis as shoreic acid (1), cabraleahydroxylactone (2), cabralealactone (3), cinchonain (5), catechin (6), scopoletin (7), vanillic acid (8), p-hydroxybenzoic acid (9), docosanol (10), beta-sitosterol (11), daucosterol (12). Of them, compounds 1-6,8-12 were separated from this plant for the first time, and compounds 4-6 were reported from this plant genus for the first time.
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Supelco
β-Sitosterol β-D-glucoside, analytical standard
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Vanillic acid, 97%
Supelco
Vanillic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Scopoletin, analytical standard
Sigma-Aldrich
Scopoletin, ≥99%