Merck
  • Enzyme kinetic and molecular docking studies on the metabolic interactions of 1-hydroxy-2,3,5-trimethoxy-xanthone, isolated from Halenia elliptica D. Don, with model probe substrates of human cytochrome P450 enzymes.
Enzyme kinetic and molecular docking studies on the metabolic interactions of 1-hydroxy-2,3,5-trimethoxy-xanthone, isolated from Halenia elliptica D. Don, with model probe substrates of human cytochrome P450 enzymes.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2012-07-24)
Ru Feng, Xuelin Zhou, Penelope M Y Or, Jing-Yi Ma, Xiang-Shan Tan, Jie Fu, Chao Ma, Jian-Gong Shi, Chun-Tao Che, Yan Wang, John H K Yeung
ABSTRACT
Halenia elliptica D. Don is a Tibetan herb and medicinal preparations containing Halenia elliptica have been commonly used for the treatment of hepatitis B virus infection in China. The metabolism of 1-hydroxy-2,3,5-trimethoxy-xanthone (HM-1) to its metabolites is mediated through cytochrome P450 enzymes. This study aimed to investigate the herb-drug interaction potential of HM-1 by studying its effects on the metabolism of model probe substrates of five major CYP450 isoforms in human liver microsomes. HM-1 showed moderate inhibitory effects on CYP1A2 (IC₅₀ = 1.06 μM) and CYP2C9 (IC₅₀ = 3.89 μM), minimal inhibition on CYP3A4 (IC₂₀ = 11.94 μM), but no inhibition on model CYP2D6 (dextromethorphan) and CYP2E1 (chlorzoxazone) probe substrates. Inhibition kinetic studies showed that the K(i) values of HM-1 on CYP1A2, CYP2C9 and CYP3A4 were 5.12 μM, 2.00 μM and 95.03 μM, respectively. HM-1 competitively inhibited testosterone 6β-hydroxylation (CYP3A4) but displayed mixed type inhibitions for phenacetin O-deethylation (CYP1A2) and tolbutamide 4-hydroxylation (CYP2C9). Molecular docking study confirmed the inhibition modes of HM-1 on these human CYP isoforms.
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)
Supelco
Phenacetin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phenacetin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tolbutamide, analytical standard