Merck
  • [Study on chemical constituents of the ethyl acetate extract from Blumea aromatica].
[Study on chemical constituents of the ethyl acetate extract from Blumea aromatica].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2012-07-25)
Ming-sheng Lan, Chao Luo, Chang-heng Tan, Lu Chen, Song Wei, Da-yuan Zhu
ABSTRACT
To study the chemical constituents of the ethyl acetate extract from Blumea aromatica. Column chromatographic techniques were used for the isolation and purification. Spectroscopic techniques were used for the identification of structures. Seven compounds were isolated from the ethyl acetate extract of 80% alcohol extract of the whole plant, and their structures were identified as xanthoxylin(1), dibutyl phthalate(2), luteolin-3',4',7-trimethyl ether(3), rubiadin(4), eriodictyol(5), kaempherol(6), luteolin 7,3'-dimethyl ether(7). All the compounds are isolated from this plant for the first time. Among them, compounds 2,4 and 7 are isolated from this genus for the first time.
Product Description
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Dibutyl phthalate, 99%
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Supelco
Rubiadin, analytical standard