Merck
  • Comparison of greenhouse and field degradation behaviour of isoprocarb, hexaflumuron and difenoconazole in Perilla frutescens.
Comparison of greenhouse and field degradation behaviour of isoprocarb, hexaflumuron and difenoconazole in Perilla frutescens.
Bulletin of environmental contamination and toxicology (2012-08-09)
Caixia Sun, Hu Zhang, Tao Tang, Mingrong Qian, Yuwei Yuan, Zhiheng Zhang
ABSTRACT
Isoprocarb, hexaflumuron and difenoconazole were used in Perilla frutescens at 600, 60 and 75 g a.i./ha respectively. High performance liquid chromatography-tandem mass spectrometry was used for residue determination because of high selectivity and simple treatment. The results showed that the half-lives of isoprocarb, hexaflumuron and difenoconazole at greenhouse condition were 0.71, 1.63 and 1.21 days respectively, and at field condition, the values were 1.13, 1.07 and 0.92 days respectively.
Product Description
Supelco
Isoprocarb, PESTANAL®, analytical standard
Supelco
Difenoconazol, PESTANAL®, analytical standard
Supelco
Hexaflumuron, PESTANAL®, analytical standard