Merck
[Chemical constituents from the fruits of Kadsura marmorata].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2012-08-11)
Hong-Jun Wang, Xin-Liang Wu, Tie-Zhong Zhou, Xu-Ming Deng, Da-Cheng Wang
ABSTRACT
To study the chemical constituents from the fruits of Kadsura marmorata. The chemical constituents were isolated and purified by chromatography on silica gel, Sephadex LH-20 column and HPLC. 9 compounds were isolated and identified as 4,5-dihydroxy-3-methoxybiphenyl (I), eriobofuran (II), 3beta, 16beta-dihydroxy urs-2-ene (III), 2alpha, 3beta, 6beta, 23-tetrahydroxy urs-12,18-dien-28-oic acid (IV), 2alpha,3beta,23-trihydroxy urs-12-en-28-oic acid (V), rutin (VI), 2-ethylhexanoic acid (VII), 2-monolaurin (VIII), glyceryl monoricinoleate (IX) on the basis of NMR and EI-MS spectroscopic analysis. All these compounds are isolated from this genus for the first time.
Sigma-Aldrich
2-Ethylhexanoic acid, 99%
Sigma-Aldrich
2-Ethylhexanoic acid, ≥99%
Sigma-Aldrich
2-Ethylhexanoic acid, ≥99%
Supelco
2-Ethylhexanoic acid, analytical standard