Merck
  • First measurements of the scope for growth (SFG) in mussels from a large scale survey in the North-Atlantic Spanish coast.
First measurements of the scope for growth (SFG) in mussels from a large scale survey in the North-Atlantic Spanish coast.
The Science of the total environment (2012-08-14)
Marina Albentosa, Lucía Viñas, Victoria Besada, Angeles Franco, Amelia González-Quijano
ABSTRACT
SFG and physiological rates were measured in wild mussels from the Spanish Marine Pollution monitoring program (SMP) in order to determine seawater quality. It consists of 41 stations, covering almost 2500 km of coast, making the SMP the widest-ranging monitoring network in the Iberian Peninsula's Atlantic region. Results of the 2007 and 2008 surveys when 39 sites were sampled: (20 in 2007 and 19 in 2008, being 8 sites sampled both years) were presented. Chemical analyses were carried out to determine the relationships between physiological rates and the accumulation of toxic compounds. Data presented are the first to become available on the use of SFG as a biomarker of the marine environment on a large spatial scale (>1000 km) along Spain's Atlantic seaboard. SFG values enable significant differences to be established between the areas sampled and between the two years surveyed. The integration of biological and chemical data suggests that certain organochlorine compounds, namely chlordanes and DDTs, may have a negative effect on SFG, although such an effect is of a lesser magnitude than that associated with certain biological parameters such as condition index and mussel age. These variables act as confounding factors when attempting to determine the effect of chemical compounds present in the marine environment on mussel SFG. Further research is therefore needed on the relation between these confounding factors and SFG in order to apply the relevant corrective strategies to enable this index to be used in monitoring programs. The effect of these confounding factors is more clearly revealed in studies that cover a wide-ranging spatial and time scale, such as those carried out within the SMP. These results do not invalidate the use of biological data in monitoring programs, but rather point to the need to analyze all the factors affecting each biological process.