Merck
  • Visual determination of Cu2+ through copper-catalysed in situ formation of Ag nanoparticles.
Visual determination of Cu2+ through copper-catalysed in situ formation of Ag nanoparticles.
The Analyst (2012-08-15)
Xun Yuan, Yi Chen
ABSTRACT
A new strategy was explored for the visual determination of Cu(2+) using copper-catalysed in situ formation of Ag nanoparticles. In this method, only common reagents were used and the pre-synthesis and modification of nanoparticles are avoided. Ag(+) can form a milk-white suspension (AgBr) with Br(-) in an aqueous solution composed of AgNO(3), cetyltrimethylammonium bromide, ascorbic acid, bovine serum albumin, and NaNO(3). The reaction will be stopped by addition of Cu(2+), accompanied by a colour change from milk-white to orange or brilliant yellow. Cu(+) (the reduction product of Cu(2+)) consumes the dissolved O(2) and prevents the O(2) from oxidizing the newly reduced Ag atoms (by ascorbic acid) back to Ag(+), facilitating the further aggregation of Ag atoms to become Ag nanoparticles. The visible colour change was shown to be specific towards Cu(2+) over most other metal ions. The limit of detection was 0.75 μM Cu(2+) by the naked eye and 0.25 μM by spectrometer. Quantitation of Cu(2+) was achieved in a linear range from 0.25 to 2.0 μM. This method was validated by measuring real water and serum samples, giving results agreeing well with the data reported and measured by inductively coupled plasma mass spectrometry. The recovery was 95.6-106% for untreated tap water and 96.0-100% for resin-pre-treated water and serum samples.
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium nitrate, ReagentPlus®, ≥99.0%