Merck
  • Acene-doped polymer films: singlet oxygen dosimetry and protein sensing.
Acene-doped polymer films: singlet oxygen dosimetry and protein sensing.
Chemical communications (Cambridge, England) (2012-08-18)
Damla Koylu, Syena Sarrafpour, Jingjing Zhang, Sanya Ramjattan, Matthew J Panzer, Samuel W Thomas
ABSTRACT
This paper describes thin films comprising acenes dispersed in a conjugated polymeric host that have a ratiometric photoluminescence response to singlet oxygen. These films also respond to irradiation of protein-bound sensitizers, which represents a solution to the problem of protein-conjugated polymer non-specific interactions.
Sigma-Aldrich
Rubrene, sublimed grade, 99.99% trace metals basis
Sigma-Aldrich
Rubrene, ≥98%
Sigma-Aldrich
Rubrene, powder