Merck
  • In vivo corrosion mechanism by elemental interdiffusion of biodegradable Mg-Ca alloy.
In vivo corrosion mechanism by elemental interdiffusion of biodegradable Mg-Ca alloy.
Journal of biomedical materials research. Part B, Applied biomaterials (2012-08-24)
Jae-Young Jung, Sang-Jun Kwon, Hyung-Seop Han, Ji-Young Lee, Jae-Pyoung Ahn, Seok-Jo Yang, Sung-Youn Cho, Pil-Ryung Cha, Yu-Chan Kim, Hyun-Kwang Seok
ABSTRACT
We elucidated the in vivo corrosion mechanism of the biodegradable alloy Mg-10 wt % Ca in rat femoral condyle through transmission electron microscope observations assisted by focused ion beam technique. The alloy consists of a primary Mg phase and a three-dimensional lamellar network of Mg and Mg(2)Ca. We found that the Mg(2)Ca is rapidly corroded by interdiffusion of Ca and O, leading to a structural change from lamellar network to nanocrystalline MgO. In contrast to the fast corrosion rate of the lamellar structure, the primary Mg phase slowly changes into nanocrystalline MgO through surface corrosion by O supplied along the lamellar networks. The rapid interdiffusion induces an inhomogeneous Ca distribution and interestingly leads to the formation of a transient CaO phase, which acts as a selective leaching path for Ca. In addition, the outgoing Ca with P from body fluids forms needle-type calcium phosphates similar to hydroxyl apatite at interior and surface of the implant, providing an active biological environment for bone mineralization.
Sigma-Aldrich
Magnesium oxide, BioUltra, ≥97.0% (calcined substance, KT)
Sigma-Aldrich
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
Sigma-Aldrich
Magnesium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, 99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, tested according to Ph. Eur., heavy
Sigma-Aldrich
Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Magnesium oxide, -10-+50 mesh, 98%
Sigma-Aldrich
Magnesium oxide, light, 95%
Sigma-Aldrich
Magnesium oxide, ACS reagent, 97%
Sigma-Aldrich
Magnesium oxide, ≥99% trace metals basis, -325 mesh