Merck
  • The impact of rotigotine on cardiovascular autonomic function in early Parkinson's disease.
The impact of rotigotine on cardiovascular autonomic function in early Parkinson's disease.
European neurology (2012-09-06)
Camilla Rocchi, Mariangela Pierantozzi, Valerio Pisani, Girolama Alessandra Marfia, Alessandra Di Giorgio, Paolo Stanzione, Giorgio Bernardi, Alessandro Stefani
ABSTRACT
Dysautonomia can occur in early stages of Parkinson's disease (PD) influencing tolerance to dopaminergic therapies. Rotigotine, a non-ergot dopamine agonist, has recently been developed as an effective alternative antiparkinsonian drug, but its influence on the autonomic nervous system was not investigated. Twenty subjects out of 34 consecutive de novo PD patients were submitted to full assessment of cardiovascular autonomic function before and after reaching a stable rotigotine regimen [6 mg/24 h (n = 3) or 8 mg/24 h (n = 17)]. Patients reached significant clinical improvement (-27% on the Unified Parkinson's Disease Rating Scale part III) and did not show significant differences in cardiovascular tests compared to baseline data. However, an unexpected trend towards increasing systolic blood pressure after head-up tilt test was detected. Our study demonstrates that rotigotine does not influence cardiovascular autonomic responses in early de novo PD patients. Consequently, it may represent a well-tolerated and efficacious therapeutic option in newly diagnosed PD subjects.
Sigma-Aldrich
Rotigotine hydrochloride, ≥98% (HPLC)