Merck
  • Effects of addition of egg ovotransferrin-derived peptides on the oxygen radical absorbance capacity of different teas.
Effects of addition of egg ovotransferrin-derived peptides on the oxygen radical absorbance capacity of different teas.
Food chemistry (2012-09-08)
Wuyang Huang, Shengwen Shen, Chamila Nimalaratne, Sen Li, Kaustav Majumder, Jianping Wu
ABSTRACT
Ovotransferrin-derived peptides showed synergistic effects with vitamin C, epigallocatechin gallate (EGCG), and caffeic acid, but not quercetin in our previous report. In this study, we further investigated the interactions between ovotransferrin-derived peptides and teas, based on the oxygen radical absorbance capacity (ORAC) assay. Our results showed that there was no significant difference in ORAC values among green, oolong, and black teas. For all the samples, 80% methanol extracts possessed better antioxidant capacity than hot water extracts. The antioxidant capacity of teas were improved by adding either ovotransferrin hydrolysate or its purified peptide IRW; however, adding hydrolysate did not improve antioxidant stability of teas. ORAC values of both teas and hydrolysate added teas were decreased during 22weeks of storage, while samples stored at 4°C exhibited higher antioxidant capacity than those stored at room temperature. This suggested that ovotransferrin hydrolysate could be used as functional food ingredients in enhancing antioxidant capacities of foods, which would benefit human nutrition and health.
Sigma-Aldrich
Conalbumin from chicken egg white, Iron complex
Sigma-Aldrich
Conalbumin from chicken egg white, suitable for cell culture, BioReagent
Sigma-Aldrich
Conalbumin from chicken egg white, Substantially iron-free