Merck
  • A framework for identifying characteristic odor compounds in municipal wastewater effluent.
A framework for identifying characteristic odor compounds in municipal wastewater effluent.
Water research (2012-09-18)
Eva Agus, Lifeng Zhang, David L Sedlak
ABSTRACT
Municipal wastewater often contains trace amounts of organic compounds that can compromise aesthetics of drinking water and undermine public confidence if a small amount of effluent enters the raw water source of a potable water supply. To efficiently identify compounds responsible for odors in wastewater effluent, an analytical framework consisting of gas chromatography with mass spectrometry (GC-MS) and gas chromatography with olfactometry detection (GC-Olf) coupled with flavor profile analysis (FPA) was used to identify and monitor compounds that could affect the aesthetics of drinking water. After prioritizing odor peaks detected in wastewater effluent by GC-Olf, the odorous components were tentatively identified using retention indices, mass spectra and odor descriptors. Wastewater effluent samples were typically dominated by earthy-musty odors with additional odors in the amine, sulfidic and fragrant categories. 2,4,6-trichloroanisole (246TCA), geosmin and 2-methylisoborneol (2MIB) were the main sources of the earthy/musty odors in wastewater effluent. The other odors were attributable to a suite of compounds, which were detected in some but not all of the wastewater effluents at levels well in excess of their odor thresholds. In most cases, the identities of odorants were confirmed using authentic standards. The fate of these odorous compounds, including 2-pyrrolidone, methylnaphthalenes, vanillin and 5-hydroxyvanillin (5-OH-vanillin), should be considered in future studies of water systems that receive effluent from upstream sources.
Sigma-Aldrich
2-Methylisoborneol solution, ~10 mg/mL in methanol, ≥98% (GC)
Sigma-Aldrich
2-Methylisoborneol, ≥98.0% (GC)