Merck
  • UV-H2O2 degradation of methyl orange catalysed by H3PW12O40/activated clay.
UV-H2O2 degradation of methyl orange catalysed by H3PW12O40/activated clay.
Environmental technology (2012-09-20)
Guangtao Wei, Linye Zhang, Tengyou Wei, Qiyu Luo, Zhangfa Tong
ABSTRACT
A catalyst consisting of phosphotungstic acid (H3PW12O40) combined with activated clay was prepared by the impregnation method, and an experiment was carried out to evaluate the catalytic activity of the H3PW12O40/activated clay for the degradation of methyl orange (MO) in the UV-H2O2 process. The degradation ratio of MO can be affected by H2O2 concentration, reaction time, catalyst dosage, pH and temperature. The reaction temperature should be controlled at less than 70 degrees C, and the catalyst has a wide applicable pH range in the UV-H2O2 process. Hydroxyl radicals were generated in the UV-H2O2 system under the action of H3PW12O40/activated clay, and MO was degraded by hydroxyl radicals. Compared with traditional catalysts used in UV-H2O2 systems, H3PW12O40/activated clay has certain advantages for its practical application.
Sigma-Aldrich
Methyl Orange, suitable for microscopy
Sigma-Aldrich
Methyl Orange, ACS reagent, Dye content 85 %