Merck
  • Physicochemical and thermal characteristics of the sludge produced after thermochemical treatment of petrochemical wastewater.
Physicochemical and thermal characteristics of the sludge produced after thermochemical treatment of petrochemical wastewater.
Environmental technology (2012-09-20)
Shilpi Verma, Basheshwar Prasad, I M Mishra
ABSTRACT
The present work describes the physicochemical and thermal characteristics of the sludge generated after thermochemical treatment of wastewater from a petrochemical plant manufacturing purified terephthalic acid (PTA). Although FeCl3 was found to be more effective than CuSO4 in removing COD from wastewater, the settling and filtration characteristics of FeCl3 sludge were poorer. Addition of cationic polyacrylamide (CPAA; 0.050kg/m3) to the FeCl3 wastewater system greatly improved the values of the filter characteristics of specific cake resistance (1.2 x 10(8) m/kg) and resistance of filter medium (9.9 x 10(8) m(-1)) from the earlier values of 1.9 x 10(9) m/kg and 1.7 x 10(8) m(-1), respectively. SEM-EDAX and FTIR studies were undertaken, to understand the sludge structure and composition, respectively. The moisture distribution in the CuSO4 sludge, FeCl3 sludge and FeCl3 + CPAA sludge showed that the amount of bound water content in the CuSO4 and FeCl3 + CPAA sludges is less than that of the FeCl3 sludge and there was a significant reduction in the solid-water bond strength of FeCl3 + CPAA sludge, which was responsible for better settling and filtration characteristics. Due to the hazardous nature of the sludge, land application is not a possible route of disposal. The thermal degradation behaviour of the sludge was studied for its possible use as a co-fuel. The studies showed that degradation behaviour of the sludge was exothermic in nature. Because of the exothermic nature of the sludge, it can be used in making fuel briquettes or it can be disposed of via wet air oxidation.
Sigma-Aldrich
Copper(II) sulfate, ReagentPlus®, ≥99%
Sigma-Aldrich
Copper(II) sulfate solution, 4 % (w/v) (prepared from copper (II) sulfate pentahydrate)
Sigma-Aldrich
Copper(II) sulfate, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Terephthalic acid, 98%
Sigma-Aldrich
Iron(II) chloride, 98%
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Iron(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Polyacrylamide, nonionic water-soluble polymer
Sigma-Aldrich
Iron(II) chloride tetrahydrate, ReagentPlus®, 98%
Sigma-Aldrich
Iron(II) chloride tetrahydrate, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Polyacrylamide, average Mn 150,000