Merck
  • Macromolecular and morphological evolution of poly(styrene sulfonate) complexes with tetradecyltrimethylammonium bromide.
Macromolecular and morphological evolution of poly(styrene sulfonate) complexes with tetradecyltrimethylammonium bromide.
The journal of physical chemistry. B (2012-09-21)
Alexey Popov, Julia Zakharova, Alexander Wasserman, Mikhail Motyakin, Victor Kasaikin
ABSTRACT
Macromolecular characteristics and morphology of water-soluble complexes between sodium poly(styrene sulfonate) (PSS) and tetradecyltrimethylammonium bromide have been followed as a function of surfactant-to-polymer charge ratio (S/P) to elicit possible changes in the complexation mechanism. As revealed by light scattering, shorter PSS (30 and 150 repeat units) yield multichain complexes while longer PSS (450 and 5000 repeat units) form single-chain species throughout 0 < S/P < 0.9. Irrespective of PSS chain length, the complexes exist in solution in a swollen coil conformation and undergo a compaction with S/P but never collapse into a globule. Even when the free PSS chain is too short to coil (30 repeat units), the complexes adopt a coiled conformation due to multichain aggregation. Morphological changes (manifested by a hypochromic shift in UV spectra of the complexes at S/P < 0.5 and an increase in the local surfactant mobility observed at S/P > 0.5 by ESR) strongly suggest a change in the formation mechanism of the complexes with a transition near S/P = 0.5.
Sigma-Aldrich
Poly(4-styrenesulfonic acid) solution, Mw ~75,000, 18 wt. % in H2O
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate), average Mw ~70,000
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate), average Mw ~1,000,000, powder
Supelco
Myristyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate) solution, average Mw ~70,000, 30 wt. % in H2O
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate) solution, average Mw ~1,000,000, 25 wt. % in H2O
Sigma-Aldrich
Myristyltrimethylammonium bromide, ≥99%
Sigma-Aldrich
Trimethyl-tetradecylammonium chloride, ≥98.0% (AT)
Sigma-Aldrich
Myristyltrimethylammonium bromide, 98% (AT)
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate) solution, average Mw ~200,000, 30 wt. % in H2O