Merck
  • White adipose tissue apolipoprotein C-I secretion in relation to delayed plasma clearance of dietary fat in humans.
White adipose tissue apolipoprotein C-I secretion in relation to delayed plasma clearance of dietary fat in humans.
Arteriosclerosis, thrombosis, and vascular biology (2012-09-22)
Hanny Wassef, Huda Salem, Simon Bissonnette, Alexis Baass, Robert Dufour, Jean Davignon, May Faraj
ABSTRACT
White adipose tissue (WAT) dysfunction is characterized by delayed clearance of dietary triglyceride-rich lipoproteins (TRL). We reported that apolipoprotein (apo) C-I, a transferable apolipoprotein that inhibits lipoprotein lipase activity when bound to TRL, was produced by a human adipocyte model. Thus, we aimed to determine whether increased WAT apoC-I secretion is related to delayed dietary fat clearance in humans. After the ingestion of a (13)C-triolein-labeled high-fat meal, postmenopausal obese women with high-fasting WAT apoC-I secretion (median >0.81 μmol/L per g/4 hours, n=9) had delayed postprandial plasma clearance of (13)C-triglyceride and (13)C-nonesterified fatty acids over 6 hours compared with controls. WAT apoC-I secretion over 4 hours correlated with fasting total and non-high-density lipoprotein apoC-I but not with high-density lipoprotein apoC-I and was the primary predictor of 4-hour postprandial increases in TRL apoC-I. Correction for TRL apoC-I eliminated the association of WAT apoC-I with 6-hour area under the curve of plasma (13)C-triglyceride; correction for insulin sensitivity or inflammation did not. Finally, in addition to apoC-I, WAT secreted considerable amount of apoC-II, apoC-III, and apoE over 24 hours; however, only WAT apoC-I secretion was associated with 6-hour area under the curve of plasma (13)C-triglyceride. Increased WAT apoC-I secretion in obese women is associated with delayed postprandial dietary fat clearance mediated by increased TRL apoC-I. Thus, we hypothesize that reducing WAT apoC-I secretion ameliorates WAT dysfunction and associated cardiometabolic risks in humans.
Sigma-Aldrich
Glyceryl trioleate, ≥99%
Sigma-Aldrich
Glyceryl trioleate, ~65%
Sigma-Aldrich
Glyceryl trioleate, ≥97.0% (TLC)