Merck
  • Novel CeO2 yolk-shell structures loaded with tiny Au nanoparticles for superior catalytic reduction of p-nitrophenol.
Novel CeO2 yolk-shell structures loaded with tiny Au nanoparticles for superior catalytic reduction of p-nitrophenol.
Nanoscale (2012-10-02)
Cong-Min Fan, Lin-Fei Zhang, Sha-Sha Wang, Dong-Hong Wang, Li-Qiang Lu, An-Wu Xu
ABSTRACT
Direct fabrication of core-shell or yolk-shell functional nanomaterials via a facile template-free method remains a challenge. In this work, we present a novel approach that involves straightforward chemical transformation and thermal treatment of the infinite coordination polymer particles to obtain composition-tunable CeO(2) yolk-shell structures. Uniform CeO(2) yolk-shell hollow spheres with a high surface area are promising support materials for tiny gold nanoparticles (ca. 4 nm), forming Au-CeO(2) nanocomposites which exhibit a remarkable catalytic activity and high stability for the reduction of p-nitrophenol. A possible mechanism for the formation of CeO(2) yolk-shell microspheres is also proposed.
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Supelco
4-Nitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade