Merck
  • Designing distance dependent SERS assay for monitoring photothermal antibacterial activity response.
Designing distance dependent SERS assay for monitoring photothermal antibacterial activity response.
Chemical communications (Cambridge, England) (2012-10-11)
Sadia Afrin Khan, Anant Kumar Singh, Zhen Fan, Dulal Senapati, Paresh Chandra Ray
ABSTRACT
This communication reports, for the first time, the design of distance dependent SERS assay for monitoring the photothermal antibacterial activity process.
Sigma-Aldrich
Rhodamine 6G, Dye content ~95 %
Sigma-Aldrich
Rhodamine 6G, suitable for fluorescence, BioReagent
Sigma-Aldrich
Rhodamine 6G perchlorate, Dye content 99 %
Sigma-Aldrich
Rhodamine 6G, Dye content 99 %
Supelco
Rhodamine 6G, analytical standard