Merck
  • Evidence on the formation of singlet oxygen in the donor side photoinhibition of photosystem II: EPR spin-trapping study.
Evidence on the formation of singlet oxygen in the donor side photoinhibition of photosystem II: EPR spin-trapping study.
PloS one (2012-10-11)
Deepak Kumar Yadav, Pavel Pospíšil
ABSTRACT
When photosystem II (PSII) is exposed to excess light, singlet oxygen ((1)O(2)) formed by the interaction of molecular oxygen with triplet chlorophyll. Triplet chlorophyll is formed by the charge recombination of triplet radical pair (3)[P680(•+)Pheo(•-)] in the acceptor-side photoinhibition of PSII. Here, we provide evidence on the formation of (1)O(2) in the donor side photoinhibition of PSII. Light-induced (1)O(2) production in Tris-treated PSII membranes was studied by electron paramagnetic resonance (EPR) spin-trapping spectroscopy, as monitored by TEMPONE EPR signal. Light-induced formation of carbon-centered radicals (R(•)) was observed by POBN-R adduct EPR signal. Increased oxidation of organic molecules at high pH enhanced the formation of TEMPONE and POBN-R adduct EPR signals in Tris-treated PSII membranes. Interestingly, the scavenging of R(•) by propyl gallate significantly suppressed (1)O(2). Based on our results, it is concluded that (1)O(2) formation correlates with R(•) formation on the donor side of PSII due to oxidation of organic molecules (lipids and proteins) by long-lived P680(•+)/TyrZ(•). It is proposed here that the Russell mechanism for the recombination of two peroxyl radicals formed by the interaction of R(•) with molecular oxygen is a plausible mechanism for (1)O(2) formation in the donor side photoinhibition of PSII.
Product Description
Supelco
Propyl gallate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Propyl gallate, powder
Sigma-Aldrich
Propyl gallate, ≥98.0% (HPLC)
Sigma-Aldrich
Propyl gallate, ≥98%, FCC
Sigma-Aldrich
Propyl gallate, antioxidant, ≥98.0% (HPLC)