Merck
  • Enzyme kinetic and molecular docking studies for the inhibitions of miltirone on major human cytochrome P450 isozymes.
Enzyme kinetic and molecular docking studies for the inhibitions of miltirone on major human cytochrome P450 isozymes.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2012-10-30)
Xuelin Zhou, Yan Wang, Tao Hu, Penelope M Y Or, John Wong, Yiu Wa Kwan, David C C Wan, Pui Man Hoi, Paul B S Lai, John H K Yeung
ABSTRACT
Previous studies have shown that major tanshinones isolated from Danshen (Salvia miltiorrhiza) inhibited human and rat CYP450 enzymes-mediated metabolism of model probe substrates, with potential in causing herb-drug interactions. Miltirone, another abietane type-diterpene quinone isolated from Danshen, has been reported for its anti-oxidative, anxiolytic and anti-cancer effects. The aim of this study was to study the effect of miltirone on the metabolism of model probe substrates of CYP1A2, 2C9, 2D6 and 3A4 in pooled human liver microsomes. Miltirone showed moderate inhibition on CYP1A2 (IC(50)=1.73 μM) and CYP2C9 (IC(50)=8.61 μM), and weak inhibition on CYP2D6 (IC(50)=30.20 μM) and CYP3A4 (IC(50)=33.88 μM). Enzyme kinetic studies showed that miltirone competitively inhibited CYP2C9 (K(i)=1.48 μM), and displayed mixed type inhibitions on CYP1A2, CYP2D6 and CYP3A4 with K(i) values of 3.17 μM, 24.25 μM and 35.09 μM, respectively. Molecular docking study further confirmed the ligand-binding conformations of miltirone in the active sites of these human CYP450 isoforms, and provided some information on structure-activity relationships for the CYPs inhibition by tanshinones. Taken together, CYPs inhibitions of miltirone were weaker than dihydrotanshinone, but stronger than cryptotanshinone, tanshinone I and tanshinone IIA.
Product Description
Supelco
Dextromethorphan HBr, certified reference material, pharmaceutical secondary standard
Supelco
Tolbutamide, analytical standard
Sigma-Aldrich
Dextromethorphan hydrobromide, meets USP testing specifications
Supelco
Dextromethorphan solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Dextromethorphan hydrobromide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dextromethorphan hydrobromide monohydrate, ≥99% (TLC)
Supelco
Phenacetin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phenacetin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)