Merck
  • Blockade of the renin-angiotensin system improves the early stages of liver regeneration and liver function.
Blockade of the renin-angiotensin system improves the early stages of liver regeneration and liver function.
The Journal of surgical research (2012-11-01)
Shir Lin Koh, Eleanor Ager, Cathy Malcontenti-Wilson, Vijayaragavan Muralidharan, Christopher Christophi
ABSTRACT
Partial hepatectomy is the preferred option for selected patients with colorectal cancer liver metastases (CRCLM). Sufficient liver regeneration (LR) is essential for a successful outcome in these patients. The blockade of the renin-angiotensin system (RAS) reduces the growth of several tumor types. The RAS also acts as a regulator of liver fibrosis and potentially LR. The angiotensin-converting enzyme (ACE) inhibitor, captopril, significantly inhibits the growth of CRCLM, but its effect on LR remains undefined. After 70% of partial hepatectomy, mice were randomly assigned to control or captopril-treated groups. LR was measured by liver-to-body weight ratio on days 1, 2, 4, 6, and 8. Hepatocyte proliferation, apoptosis and cell size, hepatic stellate cell (HSC) count, and sinusoidal endothelial cell density were quantified. Matrix metalloproteinase 9 (MMP-9) protein levels, liver injury markers, and RAS messenger RNA levels were also determined. At day 2, captopril increased liver-to-body weight ratio (56.5 ± 1.7 captopril versus 49.3 ± 2.4 control, P = 0.027). This was associated with increased HSC count (65.4 ± 4.8 cells per 100,000 μm(2), 48.7 ± 2.3, P = 0.007) and MMP-9 levels (0.68 ± 0.12 AU, 0.12 ± 0.04, P = 0.014). The messenger RNA levels of angiotensin-converting enzyme (P = 0.045) and angiotensin 1 receptor (P = 0.039) were reduced by captopril at day 2. Captopril enhanced early LR. This effect was associated with increased HSC numbers and MMP-9 protein, whereas hepatocyte proliferation was lower than controls. Captopril may provide a beneficial treatment option for the management of patients with CRCLM.
Sigma-Aldrich
Captopril, ≥98% (HPLC), powder
Supelco
Captopril, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Captopril, meets USP testing specifications