Merck
  • Involvement of basal and calcium-activated protein kinase C in neurotransmitter secretion in mouse motor synapses.
Involvement of basal and calcium-activated protein kinase C in neurotransmitter secretion in mouse motor synapses.
Bulletin of experimental biology and medicine (2012-11-01)
A E Gaydukov, A A Marchenkova, O P Balezina
ABSTRACT
Blocker of presynaptic protein kinase C isoforms, GF109203X, reduced quantal content of single and rhythmic evoked end-plate potentials. The increase in quantal content of single potentials under the effect of 4- aminopyridine was neutralized by 75% under the effect of L-type Ca(2+)-channel blocker nitrendipine and completely returned to the control level after protein kinase C inhibition with chelerythrine. Neither nitrendipine, nor GF109203X affected the potentiating effect of tetraethylammonium on quantal content of end-plate potentials. Thus, we discovered basal activity of presynaptic protein kinase C under normal conditions that is aimed at the maintenance of quantal content of evoked release. It has been concluded that 4-aminipyridine, but not tetraethylammonium, triggers Ca(2+) entry into the terminal, which activates protein kinase C and enhanced the evoked acetylcholine release.
Sigma-Aldrich
Nitrendipine, >95%, powder
Sigma-Aldrich
Chelerythrine chloride, ≥95% (TLC), powder
Supelco
Tetraethylammonium hydroxide solution, ~1.0 M (CH3CH2)4NOH in H2O, electrochemical grade
Sigma-Aldrich
Tetraethylammonium bromide, reagent grade, 98%
Sigma-Aldrich
Tetraethylammonium hydroxide solution, 35 wt. % in H2O
Sigma-Aldrich
Tetraethylammonium chloride, ≥98% (titration)
Supelco
Tetraethylammonium chloride, for electrochemical analysis, ≥99.0%
Sigma-Aldrich
Tetraethylammonium hydroxide solution, ~25% in methanol (~1.5 M)
Sigma-Aldrich
Tetraethylammonium chloride, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Tetraethylammonium bromide, ReagentPlus®, 99%
Sigma-Aldrich
Tetraethylammonium hydroxide solution, 20 wt. % in H2O
Sigma-Aldrich
GF 109203X, synthetic, ≥90% (HPLC)
Sigma-Aldrich
Tetraethylammonium chloride hydrate
Sigma-Aldrich
Tetraethylammonium iodide, 98%