Merck
  • Carmine red (E-120)-induced occupational respiratory allergy in a screen-printing worker: a case report.
Carmine red (E-120)-induced occupational respiratory allergy in a screen-printing worker: a case report.
B-ENT (2012-11-02)
C E Cox, D G Ebo
ABSTRACT
Here we present a case report of a patient suffering from occupational rhinoconjunctivitis and asthma due to IgE-mediated carmine red allergy. This is the first description of carmine red allergy in a screen-printing worker in which the diagnosis was documented by quantification of specific IgE antibodies, by skin tests, by a flow-assisted basophil activation test, and by a carmine red challenge test.
Sigma-Aldrich
Carminic acid, ≥90% (HPLC)
Supelco
Carminic acid, analytical standard