Merck
  • Synthesis, antioxidant and antimicrobial activity of novel vanillin derived piperidin-4-one oxime esters: preponderant role of the phenyl ester substituents on the piperidin-4-one oxime core.
Synthesis, antioxidant and antimicrobial activity of novel vanillin derived piperidin-4-one oxime esters: preponderant role of the phenyl ester substituents on the piperidin-4-one oxime core.
Bioorganic & medicinal chemistry letters (2012-11-03)
Salakatte Thammaiah Harini, Honnaiah Vijay Kumar, Javarappa Rangaswamy, Nagaraja Naik
ABSTRACT
The study has been achieved the efficient synthesis of vanillin derived piperidin-4-one oxime esters (5a-m) via four step reaction involved Mannich reaction of vanillin, acetone and ammonium acetate to obtain 2,6-bis(4-hydroxy-3-methoxyphenyl)-piperidin-4-one 2 followed by N-methylation and oximation. Further, to enhance the biological activity of vanillin derived piperidin-4-one oxime core, esterification of 4 with substituted benzoyl chlorides in the presence of strong organic base t-BuOK accomplished a series of vanillin derived piperidin-4-one oxime esters (5a-m). The synthesized analogues are screened for their antioxidant and antimicrobial studies and the preponderant effect of the phenyl ester substituents on the biological activity of piperidin-4-one oxime core was demonstrated. Among the tested compounds, 5i and 5j are emerged as outperformed antioxidants than standard Butylated hydroxy anisole (BHA) whereas, compounds 5b and 5d manifested potent antibacterial and antifungal activity than standard streptomycin and fluconazole respectively.
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Supelco
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Supelco
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Vanillin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG