Merck
  • Poly(vinylidene fluoride)/nickel nanocomposites from semicrystalline block copolymer precursors.
Poly(vinylidene fluoride)/nickel nanocomposites from semicrystalline block copolymer precursors.
Nanoscale (2012-11-10)
Vincent S D Voet, Martijn Tichelaar, Stefania Tanase, Marjo C Mittelmeijer-Hazeleger, Gerrit ten Brinke, Katja Loos
ABSTRACT
The fabrication of nanoporous poly(vinylidene fluoride) (PVDF) and PVDF/nickel nanocomposites from semicrystalline block copolymer precursors is reported. Polystyrene-block-poly(vinylidene fluoride)-block-polystyrene (PS-b-PVDF-b-PS) is prepared through functional benzoyl peroxide initiated polymerization of VDF, followed by atom transfer radical polymerization (ATRP) of styrene. The crystallization of PVDF plays a dominant role in the formation of the block copolymer structure, resulting in a spherulitic superstructure with an internal crystalline-amorphous lamellar nanostructure. The block copolymer promotes the formation of the ferroelectric β-polymorph of PVDF. Selective etching of the amorphous regions with nitric acid leads to nanoporous PVDF, which functions as a template for the generation of PVDF/Ni nanocomposites. The lamellar nanostructure and the β-crystalline phase are conserved during the etching procedure and electroless nickel deposition.
Sigma-Aldrich
Poly(vinylidene fluoride), average Mw ~275,000 by GPC, average Mn ~107,000, pellets
Sigma-Aldrich
Poly(vinylidene fluoride), average Mw ~180,000 by GPC, average Mn ~71,000, beads or pellets
Sigma-Aldrich
Poly(vinylidene fluoride), powder
Sigma-Aldrich
Poly(vinylidene fluoride), average Mw ~530,000, pellets