Merck
  • Chromatographic retention parameters in correlation analysis with in silico biological descriptors of a novel series of N-phenyl-3-methyl succinimide derivatives.
Chromatographic retention parameters in correlation analysis with in silico biological descriptors of a novel series of N-phenyl-3-methyl succinimide derivatives.
Journal of pharmaceutical and biomedical analysis (2012-11-14)
Nada Perisic-Janjic, Roman Kaliszan, Natasa Milosevic, Gordana Uscumlic, Nebojsa Banjac
ABSTRACT
Reversed-phase thin-layer chromatographic (RP TLC) retention coefficients for a newly designed series of N-phenyl-3-methyl succinimide derivatives, of a rationally expected anticonvusant activity, were determined as parameters of their lipophilicity. Basic pharmacokinetic descriptors of the agents were calculated in silico with the use of the established medicinal chemistry/drug design software. Highly significant, predictive relationships were found between the chromatographic retention constants and the bioactivity descriptors, which are assumed to account for drug absorption, distribution, elimination and toxicity (ADMETox) in humans. Among the agents investigated, the compounds with halogen substituent (Compounds nos. 9-13 in Fig. 1), were identified as the best drug candidates, because of their predicted proper pharmacokinetics, and have been selected for further research and development studies on new antiepileptic drugs. At the same time, among the congeners studied these can be indicated, which should not be rationally subjected to bioactivity tests.
Sigma-Aldrich
Succinimide
Pricing and availability is not currently available.