Merck
  • Grape seed extract as a potential remineralizing agent: a comparative in vitro study.
Grape seed extract as a potential remineralizing agent: a comparative in vitro study.
The journal of contemporary dental practice (2012-11-16)
Shiny Benjamin, Roshni Sharma, Sabeena Susan Thomas, Mohan Thomas Nainan
ABSTRACT
Remineralization is an effective treatment that may stop or reverse early tooth decay. Grape seed extract (GSE) is the potential remineralizing agent under investigation. Sound human tooth sections were obtained from the cervical portion of the root and stored in demineralizing solution at 37°C for 96 hours to induce artificial root caries lesions. The sections were divided into four treatment groups including 6.5% grape seed extract, sodium monofluorophosphate (220 ppm) with 0.05% calcium glycerophosphate, 0.5% calcium glycerophosphate and control (no treatment). An in vitro pH cycling model was used to cycle the demineralized specimens through treatment solutions, acidic buffer and neutral buffer for 8 days at 6 cycles per day. Subsequently, they were evaluated using confocal laser scanning microscope. Data were analyzed using analysis of variance (p < 0.05). GSE revealed less demineralization and more remineralization compared with other groups. GSE promotes remineralization of artificial root caries lesions. The search for the perfect remineralizing agent continues to this day. GSE could be a welcome addition to the remineralization armamentarium.
Sigma-Aldrich
Sodium fluorophosphate, 95%