Merck
  • Heme oxygenase-1 (HO-1) upregulation delays morphological and oxidative damage induced in an excitotoxic/pro-oxidant model in the rat striatum.
Heme oxygenase-1 (HO-1) upregulation delays morphological and oxidative damage induced in an excitotoxic/pro-oxidant model in the rat striatum.
Neuroscience (2012-12-04)
A L Colín-González, M Orozco-Ibarra, M E Chánez-Cárdenas, E Rangel-López, A Santamaría, J Pedraza-Chaverri, D Barrera-Oviedo, P D Maldonado
ABSTRACT
Quinolinic acid (QA)-induced overactivation of N-methyl-d-aspartate receptors yields excitotoxicity, oxidative stress and mitochondrial dysfunction, which altogether contribute to trigger a wide variety of toxic pathways with biochemical, behavioral and neuropathological alterations similar to those observed in Huntington's disease. Noteworthy, in the brains of these patients, increased expression of heme oxygenase-1 (HO-1) levels can be found. It has been proposed that this enzyme can exert a dual role, as it can be either protective or deleterious to the CNS. While some evidence indicates that its overexpression affords cellular anti-oxidant protection due to decreased concentrations of its pro-oxidative substrate heme group, and increased bilirubin levels, other reports established that high HO-1 expression and activity may result in a pro-oxidizing atmosphere due to a release of Fe(2+). In this work, we examined the temporal evolution of oxidative damage to proteins, HO-1 expression, immunoreactivity, total activity, and cell death after 1, 3, 5 and 7 days of an intrastriatal QA infusion (240 nmol/μl). QA was found to induce cellular degeneration, increasing carbonylated proteins and generating a transitory response in HO-1 mRNA, protein content, and immunoreactivity and activity in nerve cells. In order to study the role of HO-1 in the QA-induced cellular death, the tin protoporphyrin IX (SnPP), a well-known HO inhibitor, was administered to rats (30 μmol/kg, i.p.). The administration of SnPP to animals treated with QA inhibited the HO activation, and exacerbated the striatal cell damage induced by QA. Our findings reveal a potential modulatory role of HO-1 in the toxic paradigm evoked by QA in rats. This evidence provides a valuable tool for further approaches on HO-1 regulation in neurotoxic paradigms.
Sigma-Aldrich
2,3-Pyridinedicarboxylic acid, 99%