Merck
  • Extracellular matrix in deoxycholic acid decellularized aortic heart valves.
Extracellular matrix in deoxycholic acid decellularized aortic heart valves.
Medical science monitor : international medical journal of experimental and clinical research (2012-12-05)
Oliver Bloch, Wilhelm Erdbrügger, Wolfgang Völker, Alexander Schenk, Steffen Posner, Wolfgang Konertz, Pascal M Dohmen
ABSTRACT
Only limited information is available regarding the influence of decellularization on the extracellular matrix in heart valves. Within the extracellular matrix proteoglycans (PG) play a central role in the structural organization and physical functioning of valves and in their capability of settling with endothelial and interstitial cells partially myofibroblasts. We have therefore estimated the effects of decellularization using deoxycholic acid on the structure of the extracellular matrix and PG´s in porcine aortic valves. Cupromeronic blue was used, alone or in combination with OsO₄/thio-carbo-hydrazide/OsO₄ for electron microscopic visualization. For PG and glycosaminoglycan (GAG) investigation a papain digestion was employed in combination with photometric determination using dimethylmethylene blue. The results indicate that deoxycholic acid affects the compartmentation of the PG-associated interstitial network not significantly. Compared to controls the PG-rich network was preserved even after deoxycholic acid treatment for 48 h. In parallel to electron microscopy immune assays (ELISA) showed smooth muscle cell -actin to be reduced to 0.96% ± 0.71 and total soluble protein to 6.68% ± 2.0 (n=3) of untreated controls. Protein loss corresponded well with the observations in electron micrographs of rupture and efflux of cell content. Further signs of lysis were irregular cell contours and loss of the basement membrane. Efficient cell-lysis without disintegration or loss of integrity of the interstitial PG network can be achieved by treatment of aortic valves with deoxycholic acid for 48h. This protocol might also be suitable for clinical use to optimize conditions for growth and autologous remodelling of valves.
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Copper(II) phthalocyanine, sublimed grade, Dye content 99 %
Sigma-Aldrich
Copper(II) phthalocyanine, β-form, Dye content 90 %
Sigma-Aldrich
Copper(II) phthalocyanine, triple-sublimed grade, >99.95% trace metals basis
Sigma-Aldrich
Copper(II) phthalocyanine, Dye content >99 %