Merck
  • Synthesis and characterization of multi-alkali-metal tetraphenolates and application in ring-opening polymerization of lactide.
Synthesis and characterization of multi-alkali-metal tetraphenolates and application in ring-opening polymerization of lactide.
Inorganic chemistry (2012-12-12)
Jinjin Zhang, Chunli Jian, Yuan Gao, Lei Wang, Ning Tang, Jincai Wu
ABSTRACT
A series of alkali metal complexes supported by two bulky tetraphenols were synthesized and characterized. The reactions of α,α,α',α'-tetra(3,5-di-tert-butyl-2-hydroxyphenyl)-p-xylene(para-tetraphenol) with (n)BuLi, sodium, and KSi(NMe(2))(3) gave bimetallic complexes 1, 2, and 3, respectively. Treatments of the α,α,α',α'-tetra(3,5-di-tert-butyl-2-hydroxyphenyl)-m-xylene(meta-tetraphenol) with 2 or 4 equiv of (n)BuLi afforded complexes 4 or 5, while the reactions of meta-tetraphenol with sodium and KSi(NMe(2))(3) gave only trimetallic complexes 6 and 7 for the additional p-π interaction. Complexes 1-7 were all characterized by single-crystal X-ray diffraction techniques. In the presence of benzyl alcohol, all complexes are active catalysts for the ring-opening polymerization of L-lactide. Comparatively, bimetallic complexes 1, 2, and 3 are more efficient catalysts because of their symmetric structures, in which complex 3 presents as a rare highly active potassium catalyst for the ring-opening polymerization of lactide, leading to polymers with good molecular weight control and narrow molecular weight distributions.
Product Description
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
3,6-Dimethyl-1,4-dioxane-2,5-dione, 99%
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Supelco
Benzyl alcohol, analytical standard
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG