Merck
  • Successful rechallenge for oxaliplatin hypersensitivity reactions in patients with metastatic colorectal cancer.
Successful rechallenge for oxaliplatin hypersensitivity reactions in patients with metastatic colorectal cancer.
Anticancer research (2012-12-12)
Takako Yanai, Satoru Iwasa, Hirhonobu Hashimoto, Ken Kato, Tetsuya Hamaguchi, Yasuhide Yamada, Yasuhiro Shimada, Hiroshi Yamamoto
ABSTRACT
Discontinuation of oxaliplatin-containing regimens is sometimes necessary due to hypersensitivity reactions for which effective countermeasures have not yet been identified. We retrospectively reviewed cases in which hypersensitivity reactions developed in 623 patients treated with oxaliplatin for colorectal cancer. Clinical outcomes of patients who underwent oxaliplatin rechallenge with rechallenge protocol (STEP 1: hydrocortisone, chlorpheniramine, and/or ranitidine; STEP 2: hydrocortisone with an escalating dose to 500 mg, and/or prolonged administration time of oxaliplatin; and STEP 3: STEP 2 plus a subcutaneous injection of epinephrine) were examined. Out of 623 patients, 126 (20.2%) patients developed hypersensitivity reactions. Out of these 126 patients, 99 (78.6%) underwent oxaliplatin rechallenge. As the initial oxaliplatin rechallenge, 19 patients received subsequent treatment without the rechallenge protocol and 80 patients received oxaliplatin with the rechallenge protocol of STEP 1 (n=64), STEP 2 (n=15), and STEP 3 (n=1). The median number of oxaliplatin rechallenges was 3 (range=1 to 29). The reason for treatment discontinuation was disease progression in 55 patients (56%) and hypersensitivity reactions in 21 patients (21%). Overall hypersensitivity reactions after rechallenge were observed in 59%, with grade 3/4 in 6%. The rechallenge protocol is an effective treatment option for patients with oxaliplatin hypersensitivity reactions.
Sigma-Aldrich
Ranitidine hydrochloride, solid
Supelco
Ranitidine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material