Merck
  • Unprecedented spirocyclization of 3-methyleneindoline-2-thiones during hydrolysis of the phytoalexin cyclobrassinin.
Unprecedented spirocyclization of 3-methyleneindoline-2-thiones during hydrolysis of the phytoalexin cyclobrassinin.
Bioorganic & medicinal chemistry letters (2012-12-13)
M Soledade C Pedras, Abbas Abdoli, Paulos B Chumala, Pijus Saha, Gabriele Schatte
ABSTRACT
The phytoalexin cyclobrassinin is a plant defense that has additional importance since it inhibits brassinin hydrolase, a phytoalexin detoxifying enzyme produced by the plant pathogen Alternaria brassicicola. Hence, the 1,3-thiazino[6,5-b]indole scaffold of cyclobrassinin has great application as a lead structure to design potential inhibitors of brassinin detoxification. For this reason, it is necessary to determine whether A. brassicicola is able to transform cyclobrassinin. During this work new reactions of 1,3-thiazino[6,5-b]indoles and indoline-2-thiones and their unique [4+2] cycloaddition products were discovered and characterized.
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer