Merck
  • 5-Amino-1-naphthol, a novel 1,5-naphthalene derivative matrix suitable for matrix-assisted laser desorption/ionization in-source decay of phosphorylated peptides.
5-Amino-1-naphthol, a novel 1,5-naphthalene derivative matrix suitable for matrix-assisted laser desorption/ionization in-source decay of phosphorylated peptides.
Rapid communications in mass spectrometry : RCM (2012-12-15)
Issey Osaka, Mami Sakai, Mitsuo Takayama
ABSTRACT
Although matrix-assisted laser desorption/ionization in-source decay (MALDI-ISD) is an important method for post-translational modification (PTM) analysis, the conventional matrices, 2,5-dihydroxybenzoic acid (2,5-DHB) and 1,5-diaminonaphthalene (1,5-DAN), are poor in terms of the fragment ion yields of the phosphorylated peptides. The use of 5-amino-1-naphthol (5,1-ANL) as a novel matrix for ISD of phosphorylated peptides in MALDI time-of-flight mass spectrometry (TOFMS) is described here. We have evaluated the ion yields of ISD fragments obtained from phosphorylated peptides using three 1,5-naphthalene derivatives as MALDI-ISD matrices, i.e., 5,1-ANL, 1,5-DAN and 1,5-dihydroxynaphthalene (1,5-DHN). The signal-to-noise ratio (S/N) of c'-series ions obtained from these matrices was used to estimate their suitability for MALDI-ISD of non-modified and phosphorylated peptides. The order of the S/N values of the ISD fragments for non-modified and phosphorylated peptides were 1,5-DAN > 5,1-ANL > 1,5-DHN and 5,1-ANL > 1,5-DHN > 1,5-DAN, respectively. The newly introduced matrix 5,1-ANL gave highest ion yields of ISD fragments from mono-, di-, and tetraphosphorylated peptides, while 1,5-DAN was poor in the ISD ion yields for phosphorylated peptides.
Sigma-Aldrich
5-Amino-1-naphthol, 95%
Sigma-Aldrich
Gentisic acid sodium salt hydrate, ≥98%
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, >99.0% (HPLC)
Supelco
2-Naphthylamine, analytical standard
Supelco
1,5-Diaminonaphthalene, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure