Merck
  • Rapid and efficient synthesis of nucleoside polyphosphates and their conjugates using sulfonyl imidazolium salts.
Rapid and efficient synthesis of nucleoside polyphosphates and their conjugates using sulfonyl imidazolium salts.
Current protocols in nucleic acid chemistry (2012-12-21)
Samy Mohamady, Scott D Taylor
ABSTRACT
This unit describes a method for preparing nucleoside polyphosphates and their conjugates such as nucleoside triphosphates, symmetrical and unsymmetrical dinucleoside polyphosphates and sugar nucleotides. The protocols employ sulfonyl imidazolium salts (SnISs) as coupling reagents. These reagents are prepared in two steps in very high yield from commercially available materials and can be stored for at least 1 year at -20°C. The tetra-n-butylammonium salts of nucleoside mono-, di-, or triphosphates are reacted with the SnISs to form reactive phosphoryl imidazolium donors. These donors are reacted with the tetra-n-butylammonium salts of pyrophosphate, nucleotide mono- or diphosphates or sugar-1-phosphates to give the nucleoside polyphosphates and their conjugates in excellent yields.
Sigma-Aldrich
Tetrabutylammonium azide
Supelco
Tetrabutylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Tetrabutylammonium nitrate, 97%
Supelco
Tetrabutylammonium bisulfate solution, suitable for ion pair chromatography, LiChropur, concentrate, ampule
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 1.0 M in THF
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 75 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium chloride, ≥97.0% (NT)
Sigma-Aldrich
Tetrabutylammonium bromide solution, 50 wt. % in H2O
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic, puriss., 99% (T)
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Tetrabutylammonium bisulfate solution, ~55% in H2O
Sigma-Aldrich
Tetrabutylammonium iodide, ≥99.0% (AT)
Sigma-Aldrich
Tetrabutylammonium iodide, reagent grade, 98%
Sigma-Aldrich
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium bromide, ACS reagent, ≥98.0%
Supelco
Tetrabutylammonium iodide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Tetrabutylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium cyanide, technical, ≥80%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
Supelco
Tetrabutylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 53.5-56.5% in H2O
Sigma-Aldrich
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)
Sigma-Aldrich
Tetrabutylammonium perchlorate, ≥95.0% (T)
Supelco
Tetrabutylammonium perchlorate, for electrochemical analysis, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic solution, 1.0 M in H2O