Merck
  • Changes in the viscoelastic properties of cortical bone by selective degradation of matrix protein.
Changes in the viscoelastic properties of cortical bone by selective degradation of matrix protein.
Journal of biomechanics (2012-12-25)
Hideki Shirakawa, Kazuya Furusawa, Akimasa Fukui, Shigeru Tadano, Naoki Sasaki
ABSTRACT
We have studied stress relaxation of bovine femoral cortical bone specimens treated with KOH aqueous solution which had been known to degrade selectively protein molecules in bone. With the KOH treatment, we found an increase in specimens' volume. This increase was regarded as swelling of the bone specimen, presumably due to matrix protein network degradation including that of collagen. In an analogy of bone to gel structure, an increasing ratio of specimen volume was used as an indicating parameter for the matrix protein network degradation by the treatment. Although an empirical equation with a linearly combined form of two Kohlrausch-Williams-Watts (KWW) functions has been shown to describe the stress relaxation of bone specimens, a single KWW function was suitable for the bone specimens treated with KOH solution for as little as 3h. In KOH treated specimens, both the initial modulus and the relaxation time decreased with the volume-increasing ratio, while the relaxation time distribution did not change. A chemo-rheological consideration attributed the reduction of modulus values to the network degradation in the organic matrix phase. The relaxation time of KOH treated specimens was thought to be related to the longer relaxation time of untreated bones, although there was a discontinuity between the extrapolated relaxation time values for KOH treated specimens and untreated specimens. This discontinuity may have originated from the release of residual stress existing in the bone by the matrix protein degradation. The results of the present study suggest that the state of matrix protein is crucial for integrating the mechanical properties of bone.
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Supelco
trans-4-Hydroxy-L-proline, analytical standard
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioReagent, suitable for cell culture, ≥98.5%
Sigma-Aldrich
trans-4-Hydroxy-L-proline, ≥99%