Merck
  • Triclosan-coated sutures reduce the incidence of wound infections and the costs after colorectal surgery: a randomized controlled trial.
Triclosan-coated sutures reduce the incidence of wound infections and the costs after colorectal surgery: a randomized controlled trial.
Surgery (2012-12-25)
Toru Nakamura, Nobuichi Kashimura, Takehiro Noji, On Suzuki, Yoshiyasu Ambo, Fumitaka Nakamura, Akihiro Kishida
ABSTRACT
In colorectal surgeries, surgical site infections (SSIs) frequently cause morbidity; an incidence of up to 20% has been shown in previous studies. Recently, to prevent microbial colonization of suture material in operative wounds, triclosan-coated polyglactin suture materials with antimicrobial activity have been developed; however, their significance in colorectal surgery remains unclear. This randomized controlled trial was conducted to assess the value of triclosan-coated polyglactin sutures in colorectal surgery. A total of 410 consecutive patients who had undergone elective colorectal operations were enrolled in this trial. Of those patients, the 206 in the study group underwent wound closure with triclosan-coated polyglactin 910 antimicrobial sutures, and the 204 patients in the control group received conventional wound closures with polyglactin 910 sutures. The study group and the control group were comparable regarding risk factors for SSIs. The incidence of wound infection in the study group was 9 of 206 patients (4.3%), and that in the control group was 19 of 204 patients (9.3%). The difference is statistically significant in the 2 groups (P = .047). The median additional cost of wound infection management was $2,310. The actual entire additional cost, therefore, of 9 patients in the study group was $18,370, and that of 19 patients in the control group was $60,814. Triclosan-coated sutures can reduce the incidence of wound infections and the costs in colorectal surgery.
Sigma-Aldrich
Resomer® RG 504 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 38,000-54,000
Sigma-Aldrich
Resomer® RG 505, Poly(D,L-lactide-co-glycolide), ester terminated, Mw 54,000-69,000
Sigma-Aldrich
Resomer® RG 502 H, Poly(D,L-lactide-co-glycolide), acid terminated, viscosity 0.16-0.24 dL/g 
Sigma-Aldrich
Resomer® RG 502, Poly(D,L-Lactide-co-Glycolide), lactide:glycolide 50:50, ester terminated, Mw 7,000-17,000
Sigma-Aldrich
Resomer® RG 752 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 75:25, Mw 4,000-15,000
Sigma-Aldrich
Resomer® RG 653 H, Poly(D,L-lactide-co-glycolide), acid terminated, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 503 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 756 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 75:25, Mw 76,000-115,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide 65:35, Mw 40,000-75,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (50:50), mol wt 30,000-60,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (75:25), mol wt 66,000-107,000
Sigma-Aldrich
Triclosan, 97.0-103.0% (active substance, GC)
Sigma-Aldrich
Resomer® RG 504, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 38,000-54,000
Sigma-Aldrich
Resomer® RG 858 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 85:15, Mw 190,000-240,000
Sigma-Aldrich
Resomer® RG 503, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 24,000-38,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), ester terminated, Mw 50,000-75,000