Merck
  • Evaluation of the suspending properties of two local Opuntia spp. mucilages on paracetamol suspension.
Evaluation of the suspending properties of two local Opuntia spp. mucilages on paracetamol suspension.
Pakistan journal of pharmaceutical sciences (2012-12-25)
Naod Gebresamuel, Tsige Gebre-Mariam
ABSTRACT
Some excipients are currently available for the formulation of pharmaceutical suspensions. The purpose of this study is to develop cheap and effective natural excipient that can be used as an effective alternative for the formulation of pharmaceutical suspensions. The suspending properties of Opuntia ficus-indica and Opuntia stricta mucilages (family Cactaceae) were evaluated comparatively with that of NaCMC at concentration range of 2-6% (w/v) in Paracetamol suspension. Sedimentation volume (%) (with and without electrolyte), rheology, redispersibility, and dissolution rate of the suspensions were employed as evaluation parameters. The values obtained were used as basis for comparison of the suspending agents. The apparent viscosities of the suspensions in all the suspending agents concentration levels and applied shear rates were in the order of NaCMC>OS>OFI with non-Newtonian flow and accordingly the flow rates of the suspensions were in the order of OFI > OS > NaCMC. The sedimentation volumes (%) of the suspensions in all the suspending agent concentration levels were higher for OS followed by OFI and then NaCMC. The high sedimentation volumes (%) of suspensions, in turn, were accompanied by ease of redispersibility of that order. The effect of electrolyte on sedimentation volume (%) had dual effect. It was only the suspensions that had NaCMC that showed increase in sedimentation volume (%) in all molar NaCl concentration. However, in suspensions that had mucilages of OS and OFI, an initial increase in sediment volumes (%) were accompanied by decrease after 1x10(-3)M and 1x10(-2)M of NaCl, respectively. Dissolution of the suspensions which had mucilages attained the acceptable ranges (> 80% drug release in 30 min) in 5 min. Similarly, except A6 formulations A2, A3, A4 and A5 have attained the limit but the release was not as quick as the previous formulations. Hence, it can be concluded that mucilages of Opuntia spp. (Opuntia ficus-indica and Opuntia stricta) can be used as alternatives to NaCMC as suspending agent in suspension formulations.
Sigma-Aldrich
Carboxymethylcellulose sodium, meets USP testing specifications, Medium viscosity
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 50-200 cP , c=4% H2O at 25­°C
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 2500-6000 cP 
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.7
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 1.2
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.9
Sigma-Aldrich
Carboxymethylcellulose sodium salt, ultra high viscosity, highly purified
Sigma-Aldrich
Carboxymethylcellulose sodium salt, High viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, low viscosity