Merck
  • Monitoring of thiopurine metabolites - a high-performance liquid chromatography method for clinical use.
Monitoring of thiopurine metabolites - a high-performance liquid chromatography method for clinical use.
Journal of pharmaceutical and biomedical analysis (2012-12-25)
Svante Vikingsson, Sven Almer, Curt Peterson, Björn Carlsson, Martin Josefsson
ABSTRACT
A high-performance liquid chromatography method capable of measuring thiopurine mono-, di-, and triphosphates separately in red blood cells (RBCs) was developed. RBCs were isolated from whole blood using centrifugation. Proteins were precipitated using dichloromethane and methanol. The thioguanine nucleotides (TGNs) were derivatised using potassium permanganate before analysis. Analytes were separated by ion-pairing liquid chromatography using tetrabutylammonium ions and detected using UV absorption and fluorescence. The method was designed for use in clinical trials. Ten patient samples were analysed to demonstrate clinical application and to establish pilot ranges for all analytes. The method measured thioguanosine mono-(TGMP), di-(TGDP), and triphosphate (TGTP), as well as methylthioinosine mono- (meTIMP), di- (meTIDP) and triphosphate (meTITP) in RBCs collected from patients treated with thiopurine drugs (azathioprine, 6-mercaptopurine, and 6-thioguanine). LOQ was 0.3, 3, 2, 30, 30 and 40 pmol/8 × 10⁸ RBC, for TGMP, TGDP, TGTP, meTIMP, meTIDP and meTITP, respectively. Between-day precision were below 14% for all analytes at all concentrations and samples were stable at 4 °C for 8 h after sampling.
Sigma-Aldrich
Tetrabutylammonium hydrogensulfate, 97%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 1.0 M in methanol
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
Sigma-Aldrich
6-Mercaptopurine monohydrate, 98%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
Supelco
Tetrabutylammonium hydroxide solution, ~40% in water
Sigma-Aldrich
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 53.5-56.5% in H2O
Supelco
Tetrabutylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium perchlorate, ≥95.0% (T)
Supelco
Tetrabutylammonium perchlorate, for electrochemical analysis, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium cyanide, 95%
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic solution, 1.0 M in H2O
Sigma-Aldrich
Tetrabutylammonium cyanide, technical, ≥80%
Sigma-Aldrich
Potassium permanganate, ACS reagent, ≥99.0%, low in mercury
Sigma-Aldrich
Potassium permanganate, 97%
Sigma-Aldrich
Potassium permanganate, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium bromide, ACS reagent, ≥98.0%
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 75 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium nitrate, 97%
Sigma-Aldrich
Tetrabutylammonium bisulfate solution, ~55% in H2O
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic, puriss., 99% (T)
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 1.0 M in THF
Supelco
Tetrabutylammonium iodide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium iodide, ≥99.0% (AT)
Supelco
Tetrabutylammonium bisulfate solution, suitable for ion pair chromatography, LiChropur, concentrate, ampule
Sigma-Aldrich
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium chloride, ≥97.0% (NT)
Supelco
Tetrabutylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium bromide solution, 50 wt. % in H2O