Merck
  • Structural and photoelectrochemical evaluation of nanotextured Sn-doped AgInS(2) films prepared by spray pyrolysis.
Structural and photoelectrochemical evaluation of nanotextured Sn-doped AgInS(2) films prepared by spray pyrolysis.
ChemSusChem (2013-01-03)
Qian Cheng, Xihong Peng, Candace K Chan
ABSTRACT
Spray pyrolysis was used to prepare films of AgInS(2) (AIS) with and without Sn as an extrinsic dopant. The photoelectrochemical performance of these films was evaluated after annealing under a N(2) or S atmosphere with different amounts of the Sn dopant. DFT was used to calculate the band structure of AIS and understand the role of Sn doping in the observed properties. All AIS films were n-type, and Sn was found to increase the photocurrent and carrier concentration of AIS with an optimum doping level of x=[Sn]/([Ag]+[In])=0.02, which gave a photocurrent of 4.85 mA cm(-2). Above this level, the Sn dopants were detrimental to the photoelectrochemical performance, likely a result of a self-compensating effect and the introduction of a deep acceptor level, which could act as a recombination site for photogenerated carriers.
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Tin, ≥99%, powder
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sigma-Aldrich
Tin, 99.8%, shot, 3 mm
Sigma-Aldrich
Tin, shot, 99.999% trace metals basis
Sigma-Aldrich
Tin, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sigma-Aldrich
Tin, powder, <45 μm particle size, 99.8% trace metals basis