Merck
  • Synthesis of stable phosphomide ligands and their use in Ru-catalyzed hydrogenations of bicarbonate and related substrates.
Synthesis of stable phosphomide ligands and their use in Ru-catalyzed hydrogenations of bicarbonate and related substrates.
ChemSusChem (2013-01-03)
Saravanan Gowrisankar, Christopher Federsel, Helfried Neumann, Carolin Ziebart, Ralf Jackstell, Anke Spannenberg, Matthias Beller
ABSTRACT
New benzoyl- and naphthoyl-substituted phosphines have been synthesized, which are stable to air and moisture. Testing these so-called phosphomide ligands in the presence of different ruthenium precursors, the hydrogenation of sodium bicarbonate (NaHCO(3)) to sodium formate (NaHCO(2)) proceeded with good catalyst turnover numbers in the range of 1300-1600 at 80°C and a total pressure of hydrogen of 60 bar in the absence of amines or other additives. Similarly, catalytic hydrogenations of carbon dioxide, cinnam-, and benzaldehyde were possible with these new ruthenium complexes. As an intermediate of the catalytic cycle the defined ruthenium complex [(η(6)-C(6)H(6))-RuCl(2)(Cy(2)P(1-naphthoyl)] (Cy=cyclohexyl) was prepared and characterized by X-ray crystallography.
Product Description
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Benzaldehyde, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Benzaldehyde, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Benzaldehyde, natural, FCC, FG
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Cinnamaldehyde, natural, ≥95%, FG
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Supelco
Benzaldehyde, analytical standard
Benzaldehyde, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzaldehyde, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
trans-Cinnamaldehyde, ≥99%
Sigma-Aldrich
trans-Cinnamaldehyde, 97%
Sigma-Aldrich
trans-Cinnamaldehyde, FCC, FG
Supelco
trans-Cinnamaldehyde, analytical standard