Merck
  • Prolinethiol ether catalysis in an asymmetric Michael reaction: solvent-free synthesis of functionalized monohaloalkenes.
Prolinethiol ether catalysis in an asymmetric Michael reaction: solvent-free synthesis of functionalized monohaloalkenes.
The Journal of organic chemistry (2013-01-05)
Ai-Bao Xia, Chao Wu, Dan-Qian Xu, Yi-Feng Wang, Xiao-Hua Du, Zhao-Bo Li, Zhen-Yuan Xu
ABSTRACT
The organocatalytic Michael reaction of ketones with γ-monohalonitrodienes catalyzed by chiral prolinethiol ether under solvent-free conditions was developed. The described method represents a novel approach for accessing highly functionalized monohaloalkenes with α, β-stereocenters of up to >99% ee.
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor