Merck
  • Discrimination of isobaric Leu/Ile residues by MALDI in-source decay mass spectrometry.
Discrimination of isobaric Leu/Ile residues by MALDI in-source decay mass spectrometry.
Journal of the American Society for Mass Spectrometry (2013-01-12)
Daiki Asakawa, Nicolas Smargiasso, Edwin De Pauw
ABSTRACT
MALDI in-source decay (ISD) has been used for top-down sequencing of proteins. The use of the matrix 1,5-diaminonapthalene (1,5-DAN) gave abundant w ions, which are formed from the unimolecular dissociation of z• radical fragments via α cleavage reaction and thus help identify which of the isobaric amino acids, Leu or Ile, is present. The high abundance of w ions in MALDI-ISD with 1,5-DAN results from the low collision rate in the MALDI plume. MALDI-ISD with 1,5-DAN appears to be an useful method for the top-down sequencing of proteins, including discrimination of Leu and Ile near the C-terminal end.
Sigma-Aldrich
Gentisic acid sodium salt hydrate, ≥98%
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Anthranilic acid, reagent grade, ≥98%
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Anthranilic acid, puriss. p.a., ≥99.5% (T)
SAFC
L-Isoleucine
Supelco
1,5-Diaminonaphthalene, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, >99.0% (HPLC)
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Supelco
2-Naphthylamine, analytical standard
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%