- Specific indication of hemoproteins in polyacrylamide gels using a double-staining process.
Specific indication of hemoproteins in polyacrylamide gels using a double-staining process.
Analytical biochemistry (1984-02-01)
R T Francis, R R Becker
PMID6202169
ABSTRACT
Hemoproteins were revealed in polyacrylamide gels in the presence of sodium dodecyl sulfate by staining with different benzidine derivatives. When the protein samples were treated with either beta-mercaptoethanol or dithiothreitol, a significant decrease in peroxidase activity of the proteins possessing noncovalently bound heme led to diminished staining. However, when Coomassie blue R-250 staining followed the hemespecific stain it was observed that the hemoprotein bands stained more intensely than duplicate sample bands that had been stained only with the Coomassie blue R-250. This staining property allows the indication of hemoproteins in gels even after the peroxidase yield has been significantly depleted by reducing agents.
Product Description
Supelco
3,3′-Diaminobenzidine tetrahydrochloride hydrate, for spectrophotometric det. of Se, ≥97.5%
Pricing and availability is not currently available.