Merck
  • Oxidative addition versus substitution reactions of group 14 dialkylamino metalylenes with pentafluoropyridine.
Oxidative addition versus substitution reactions of group 14 dialkylamino metalylenes with pentafluoropyridine.
Inorganic chemistry (2013-01-25)
Prinson P Samuel, Amit Pratap Singh, Sankaranarayana Pillai Sarish, Julia Matussek, Ina Objartel, Herbert W Roesky, Dietmar Stalke
ABSTRACT
Dialkylamino compounds of group 14 elements (Si, Ge, Sn) in the +2 oxidation state supported by benzamidinate ligands were synthesized and treated with pentafluoropyridine. Two different modes of reactivity were observed, depending on the metal atom and the basicity of the substituent at the metal. Pentafluoropyridine undergoes oxidative addition reaction at the Si(II) and Ge(II) atoms whereas at the Sn(II) atom substitution of the NMe(2) group by the para fluorine of pentafluoropyridine occurs. The C-F bond activation by the lone pair of germanium is the first report of this kind. The Sn(II) fluoride obtained has an elongated Sn-F bond length and can be used as a good fluorinating agent. The compounds were characterized by multinuclear NMR spectroscopy, mass spectrometry, elemental analysis, and X-ray structural analysis. Single crystal X-ray structural analysis of the tin fluoride shows an asymmetric dimer with weak [Formula: see text] interactions.
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sigma-Aldrich
Tin, ≥99%, powder
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Germanium, powder, −100 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Germanium, powder, −100 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Tin, 99.8%, shot, 3 mm
Sigma-Aldrich
Tin, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Germanium, chips, 99.999% trace metals basis
Sigma-Aldrich
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sigma-Aldrich
Tin, shot, 99.999% trace metals basis
Sigma-Aldrich
Germanium, chips, 99.999% trace metals basis