Merck
  • Original TDAE strategy using propargylic chloride: rapid access to 1,4-diarylbut-3-ynol derivatives.
Original TDAE strategy using propargylic chloride: rapid access to 1,4-diarylbut-3-ynol derivatives.
Molecules (Basel, Switzerland) (2013-01-29)
Manon Roche, Thierry Terme, Patrice Vanelle
ABSTRACT
We report herein the first synthesis of propargylic alcohols using an organic reducing agent. Diarylbutynol derivatives are formed in moderate to good yields under mild conditions from the reaction of 1-(3-chloroprop-1-ynyl)-4-nitrobenzene with various aromatic aldehydes using tetrakis(dimethylamino)ethylene (TDAE) as reductant.
Sigma-Aldrich
Benzaldehyde, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Benzaldehyde, ReagentPlus®, ≥99%
Sigma-Aldrich
Propargyl alcohol, 99%
Supelco
Benzaldehyde, analytical standard
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Sigma-Aldrich
Benzaldehyde, natural, FCC, FG
Supelco
Benzaldehyde, Pharmaceutical Secondary Standard; Certified Reference Material
Benzaldehyde, European Pharmacopoeia (EP) Reference Standard