Merck
  • Flavone tetraglycosides and benzyl alcohol glycosides from the Mongolian medicinal plant Dracocephalum ruyschiana.
Flavone tetraglycosides and benzyl alcohol glycosides from the Mongolian medicinal plant Dracocephalum ruyschiana.
Journal of natural products (2013-01-30)
Erdenechimeg Selenge, Toshihiro Murata, Kyoko Kobayashi, Javzan Batkhuu, Fumihiko Yoshizaki
ABSTRACT
From an extract of the aerial parts of Dracocephalum ruyschiana, five new flavone tetraglycosides, five new benzyl alcohol glycosides, and 19 known compounds were isolated. The tetraglycosides contain a 7-O-β-d-glucopyranosyl-(1→2)-β-d-glucopyranosyl-(1→2)-[α-l-rhamnopyranosyl-(1→6)]-β-d-glucopyranosyl moiety. The benzyl alcohol glycosides had acyl groups on their glycosyl or aglycone moieties. The compounds were tested for antioxidant activity using DPPH. Although the new compounds were not active, phenylpropanoylquinic acid derivatives were revealed as radical scavengers in D. ruyschiana.
Product Description
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Supelco
Benzyl alcohol, analytical standard
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG