Merck
  • Severe adverse effects related to dermal exposure to a glyphosate-surfactant herbicide.
Severe adverse effects related to dermal exposure to a glyphosate-surfactant herbicide.
Clinical toxicology (Philadelphia, Pa.) (2013-01-31)
T P Mariager, P V Madsen, N E Ebbehøj, B Schmidt, A Juhl
ABSTRACT
This is a case of severe chemical burns following prolonged accidental exposure to a glyphosate-surfactant herbicide. The patient developed local swelling, bullae and exuding wounds. Neurological impairment followed affecting finger flexion and sensation with reduced nerve conduction. Imaging revealed oedema of the soft tissue and juxta-articular osteopenia, and a causal relationship to exposure is suggested.
Product Description
Supelco
Glyphosate, PESTANAL®, analytical standard
Sigma-Aldrich
N-(Phosphonomethyl)glycine, 96%
Sigma-Aldrich
N-(Phosphonomethyl)glycine, BioReagent, suitable for plant cell culture