Merck
  • Synthetic, 119Sn NMR spectroscopic, electrochemical, and reactivity study of organotin A3 corrolates including chiral and ferrocenyl derivatives.
Synthetic, 119Sn NMR spectroscopic, electrochemical, and reactivity study of organotin A3 corrolates including chiral and ferrocenyl derivatives.
Inorganic chemistry (2013-02-05)
Olga G Tsay, Byung-Kwon Kim, Tuong Loan Luu, Juhyoun Kwak, David G Churchill
ABSTRACT
Various R/Ar-functionalized tin 5,10,15-tris(pentafluorophenyl)corrolate derivatives are reported herein including the first ferrocenyltin corrolate species. The isopropyl, sec-butyl-, 2-methyl-n-butyl-, phenyl-, 2-thienyl-, and ferrocenyltin species have been prepared and characterized through (1)H, (13)C, and (119)Sn HMQC NMR spectroscopy, mass spectrometry, UV-vis and photoluminescent spectroscopy, and cyclic voltammetry studies. J(C/H-Sn) NMR spectroscopic couplings and ring-current effects (upfield shifting) were determined for the R-Sn axial hydrogen and carbon atoms. This report adds to older conceptually similar reports, by, i.e., Janson et al. (J. Am. Chem. Soc. 1969, 91, 5210) and Walker et al. (J. Am. Chem. Soc.1983, 105, 6923-6929), as discussed herein. Such NMR spectroscopic aspects are discussed for these model systems. Compound Sn-Ph bond cleavage was achieved by treatment with I(2).
Sigma-Aldrich
Tin, shot, 99.999% trace metals basis
Sigma-Aldrich
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Tin, 99.8%, shot, 3 mm
Sigma-Aldrich
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sigma-Aldrich
Tin, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Tin, ≥99%, powder
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis