Merck
  • Potent BRAF kinase inhibitors based on 2,4,5-trisubstituted imidazole with naphthyl and benzothiophene 4-substituents.
Potent BRAF kinase inhibitors based on 2,4,5-trisubstituted imidazole with naphthyl and benzothiophene 4-substituents.
Bioorganic & medicinal chemistry (2013-02-05)
Dan Niculescu-Duvaz, Ion Niculescu-Duvaz, Bart M J M Suijkerbuijk, Delphine Ménard, Alfonso Zambon, Lawrence Davies, Jean-Francois Pons, Steven Whittaker, Richard Marais, Caroline J Springer
ABSTRACT
The RAS-RAF-MEK-ERK pathway is hyperactivated in 30% of human cancers. BRAF is a serine-threonine kinase, belonging to this pathway that is mutated with high frequency in human melanoma and other cancers thus BRAF is an important therapeutic target in melanoma. We have designed inhibitors of BRAF based on 2,4,5-trisubstituted imidazoles with naphthyl and benzothiophene-4-substituents. Two compounds were discovered to be potent BRAF inhibitors: 1-(6-{2-[4-(2-dimethylamino-ethoxy)phenyl]-5-(pyridin-4-yl)-1H-imidazol-4-yl} benzo[b]thiophen-3-yl)-2,2,2-trifluoroethanol (1i) with BRAF IC(50)=190 nM and with cellular GI(50)=2100 nM, and 6-{2-[4-(2-dimethylamino-ethoxy)-phenyl]-5-pyridin-4-yl-3H-imidazol-4-yl}-naphthalen-1-ol (1q) with IC(50)=9 nM and GI(50)=220 nM.
Sigma-Aldrich
2,3-Benzofuran, 99%
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Imidazole, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Imidazole sodium derivative, technical grade
Sigma-Aldrich
Imidazole hydrochloride
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Imidazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Imidazole, Molecular Biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing